missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-O-acetyl-D-glucal, 99%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 51.0°C to 53.0°C |
Color | White |
Flash Point | >110°C |
Loss on Drying | 0.5% max. (30°C, 15 hrs) (vacuum) |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Quantity | 25 g |
Beilstein | 17, I, 111 |
Specific Rotation Condition | −15° (20°C c=2,EtOH) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'