Learn More
Tri(o-tolyl)phosphine, 98+%, Thermo Scientific Chemicals
Description
Tri(o-tolyl)phosphine is used in a ruthenium-catalyzed direct amination of alcohols. It is also used in Suzuki reaction. Further, it is used in the preparation of tri-ortho-phosphinselenide by reacting with selenium as a reagent. In addition to this, it acts as a ligand in coordination chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tri(o-tolyl)phosphine is used in a ruthenium-catalyzed direct amination of alcohols. It is also used in Suzuki reaction. Further, it is used in the preparation of tri-ortho-phosphinselenide by reacting with selenium as a reagent. In addition to this, it acts as a ligand in coordination chemistry.
Solubility
Soluble in alcohol. Slightly soluble in cold water. Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents.

Specifications
Spezifikation
Melting Point | 122°C to 127°C |
Quantity | 5 g |
Beilstein | 661212 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in alcohol. Slightly soluble in cold water. Insoluble in water. |
Formula Weight | 304.37 |
Percent Purity | ≥98% |
Chemical Name or Material | Tri (o-tolyl)phosphine |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.