Learn More
Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent
Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent,
£188.00 - £202.86
Specifications
Standard Concentration | 65 - 80, 10 - 20, 5 - 7% |
---|---|
For Use With | GC |
Hazards | Corrosive, Flammable |
Storage | 2°C to 8°C |
Standard Components | Pyridine, Hexamethyldisilazane, Chlorotrimethylsilane |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11547841
|
Thermo Scientific™
TS-48999 |
10 x 1 mL ampules |
£188.00
10mL |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11587841
|
Thermo Scientific™
TS-49001 |
50 mL |
£202.86
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent, Composed of HMDS, TMCS and high-purity pyridine, it rapidly produces TMS derivatives of polar compounds for GC analysis and biochemical synthesis. This versatile reagent is ideally suited for GC determinations of a range of compounds including sugars, alcohols, phenols, steroids, sterols, organic acids, and some amines.
- Derivatizes carbohydrates, phenols, steroids, sterols, organic acids, alcohols and some amines
- Useful for rapid production of TMS derivatives of polar compounds for gas chromatographic determination and biochemical synthesis
- Versatility, speed and ease of use of Tri-Suil HTP Reagent has made it the most widely used silylation formulation
Specifications
65 - 80, 10 - 20, 5 - 7% | |
Corrosive, Flammable | |
Pyridine, Hexamethyldisilazane, Chlorotrimethylsilane |
GC | |
2°C to 8°C |
For Research Use Only. Not for use in diagnostic procedures.