missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent

Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent.

Brand:  Thermo Scientific™ TS-49231

667.31 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11597680

  • £673.35 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent. Comprising TMSI in pyridine, this reagent silylates alcohols and phenols, organic acids, hydroxylamines, amino acids, carbohydrates, flavonoids, glycols and polyglycols, nucleotides, steroids, hydroxyl acids, barbituates, narcotics, indoles, and vitamins.

Additonal Product Information

Does not react with amines

May be used in the presence of water as long as there is enough reagent present to react with both the water and the sample

TRUSTED_SUSTAINABILITY
Specifications

Specifications

GC
65 - 80, 20 - 25%
2°C to 8°C
Pyridine, 1-(Trimethylsilyl)-1 H-imidazole
Flammable
25 mL
25 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent > 25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.