missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-Sodium Orthophosphate Dodecahydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
Melting Point | 75°C |
Color | White |
pH | 11.8 to 12 |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 380.13g/mol |
Packaging | Plastic powder jar |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- tri-Sodium orthophosphate dodecahydrate
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Tribasic sodium phosphate dodecahydrate,Trisodium phosphate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'