missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triallylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
Description
It is employed in the production of other chemicals. It is also mainly applied in organic synthesis and resin modifiers. It is used in the cross linking of high absorbent and the intermediates of ion exchange resin. It can also be employed in producing polyester activator and the evocating agent of butadiene polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the production of other chemicals. It is also mainly applied in organic synthesis and resin modifiers. It is used in the cross linking of high absorbent and the intermediates of ion exchange resin. It can also be employed in producing polyester activator and the evocating agent of butadiene polymerization.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from strong oxidizing agents and acids. Protect against electrostatic charges and keep away from ignition sources.
It is employed in the production of other chemicals. It is also mainly applied in organic synthesis and resin modifiers. It is used in the cross linking of high absorbent and the intermediates of ion exchange resin. It can also be employed in producing polyester activator and the evocating agent of butadiene polymerization.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from strong oxidizing agents and acids. Protect against electrostatic charges and keep away from ignition sources.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -70°C |
Density | 0.79 |
Boiling Point | 150°C to 151°C |
Flash Point | 31°C (87°F) |
Odor | Amine-like |
Refractive Index | 1.451 |
Quantity | 250 g |
UN Number | UN2610 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 137.23 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'