missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Triangular PTFE Stir Bars

Product Code. 11848872 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W x H):
12 x 6mm
20 x 8mm
25 x 8mm
35 x 9mm
25 x 14mm
40 x 14mm
50 x 12mm
55 x 14mm
Unit Size:
Pack of 10
Pack of 5
This item is not returnable. View return policy

Product Code. 11848872

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Particularly effective for dissolving solids and mixing sediments due to scraping-like action on the base of the container. The triangular PTFE stir bar provides stable stirring action for flat-bottom containers.

  • Stable, general-purpose magnetic stir bar
  • Particularly effective for dissolving solids and mixing sediments due to scraping-like action on the base of the container
  • Autoclavable

Specifications

Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
Dimensions (L x W x H) 50 x 12mm
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Height (Metric) 12mm
Width (Metric) 12mm
Length (Metric) 50 mm
Quantity 5/Pk.
Shape Triangle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.