missing translation for 'onlineSavingsMsg'
Learn More

Tributyl[3-(trifluoromethyl)phenyl]stannane, Tech., Thermo Scientific™

Product Code. 10049591
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 53566-38-4
Molecular Formula C19H31F3Sn
Molecular Weight (g/mol) 435.162
MDL Number MFCD01934759
InChI Key LBIOKWLHIBBLEI-UHFFFAOYSA-N
Synonym tributyl 3-trifluoromethyl phenyl stannane, tributyl-3-trifluoromethyl phenyl stannane, 3-tributylstannyl benzotrifluoride, 3-trifluoromethyl tri-n-butylstannyl benzene, tributyl-3-trifluoromethyl-phenyl-stannane, tributyl-3-trifluoromethyl phenyl-stannane
PubChem CID 2777910
IUPAC Name tributyl-[3-(trifluoromethyl)phenyl]stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=C1)C(F)(F)F

Specifications

Packaging Amber glass bottle
Quantity 1 g
Formula Weight 435.16
Percent Purity Tech.
Grade Technical
Chemical Name or Material Tributyl[3-(trifluoromethyl)phenyl]stannane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.