missing translation for 'onlineSavingsMsg'
Learn More

Tributyl phosphate, 99+%, Thermo Scientific Chemicals

Product Code. 10173990 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
10 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: According to the Federal Republic of Germany's Export Control Regulations and the German Chemical Companies Association (VCI), we must obtain a customer declaration for each customer before supplying

Product Code. 10173990

Brand: Thermo Scientific Acros 169930010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: According to the Federal Republic of Germany's Export Control Regulations and the German Chemical Companies Association (VCI), we must obtain a customer declaration for each customer before supplying

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-73-8
Molecular Formula C12H27O4P
Molecular Weight (g/mol) 266.32
MDL Number MFCD00009436
InChI Key STCOOQWBFONSKY-UHFFFAOYSA-N
Synonym tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de
PubChem CID 31357
ChEBI CHEBI:35019
IUPAC Name tributyl phosphate
SMILES CCCCOP(=O)(OCCCC)OCCCC

Specifications

Melting Point -79.0°C
Density 0.9790g/mL
Boiling Point 289.0°C
Flash Point 146°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Linear Formula [CH3(CH2)3O]3P(O)
Refractive Index 1.4230 to 1.4250
Quantity 1 L
Beilstein 01,II,397
Merck Index 15,9781
Specific Gravity 0.979
Solubility Information Solubility in water: 0.6g/100mL (20°C). Other solubilities: soluble in alcohols,ketones,ethers,benzene,,carbon disulfide and most organic solvents
Viscosity 3.5 mPa.s (25°C)
Formula Weight 266.32
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Tributyl phosphate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tributyl phosphate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.