missing translation for 'onlineSavingsMsg'
Learn More

Tributylethynylstannane, 95%

Product Code. 11396755 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
This item is not returnable. View return policy

Product Code. 11396755

Brand: Thermo Scientific Acros 389180025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 994-89-8 | C14H28Sn | 315.088 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 994-89-8
Color Colorless
Density 1.0800g/mL
Boiling Point 70.0°C (0.2 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C14H28Sn
Refractive Index 1.4750 to 1.4770
MDL Number MFCD00009420
Quantity 2.5 g
Specific Gravity 1.08
Synonym tributylstannylacetylene, ethynyltri-n-butyltin, ethynyltributylstannane, tributyl ethynyl stannane, stannane, tributylethynyl, tributylethynyl tin, ethynyltributyltin, tributylethynyltin, tributylstannylethyne, tributyl ethynyl tin
InChI Key YEMJHNYABQHWHL-UHFFFAOYSA-N
SMILES CCCC[Sn](CCCC)(CCCC)C#C
IUPAC Name tributyl(ethynyl)stannane
Molecular Weight (g/mol) 315.088
PubChem CID 621176
Formula Weight 315.07
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tributylethynylstannane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.