missing translation for 'onlineSavingsMsg'
Learn More
Tributylphenyltin, Thermo Scientific Chemicals Product Code.: 10156943

Tributylphenyltin, Thermo Scientific Chemicals

Product Code. 10156943
50 g, Glass bottle
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10156943

Brand: Thermo Scientific Acros 370230500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 960-16-7
Molecular Formula C18H32Sn
Molecular Weight (g/mol) 367.16
MDL Number MFCD00134394
InChI Key SYUVAXDZVWPKSI-UHFFFAOYSA-N
Synonym tributyl phenyl stannane, tributylphenyltin, phenyltributylstannane, stannane, tributylphenyl, phenyltributyltin, phenyltri-n-butyltin, phenyltributlytin, tributylphenyl tin, phenyl tnbutyl tin, phenyl tributyl tinShow MoreShow Less
PubChem CID 607632
IUPAC Name tributyl(phenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC=C1

Specifications

Color Colorless
Density 1.1250g/mL
Boiling Point 125.0°C to 128.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5Sn[(CH2)3CH3]3
Refractive Index 1.5150 to 1.5170
Quantity 50 g
Specific Gravity 1.125
Formula Weight 367.16
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Tributylphenyltin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0410147 Certificate TypeCertificate of Analysis Date21/01/2022 Catalog Number370230500,370230010,370230100
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tributylphenyltin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tributylphenyltin, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.