missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Acid, for Electrophoresis, Fisher Chemical™
£62.05 - £159.00
Chemical Identifiers
CAS | 76-03-9 |
---|---|
Molecular Formula | C2HCl3O2 |
Molecular Weight (g/mol) | 163.378 |
MDL Number | 4177 |
InChI Key | YNJBWRMUSHSURL-UHFFFAOYSA-N |
Synonym | trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer |
PubChem CID | 6421 |
ChEBI | CHEBI:30956 |
IUPAC Name | 2,2,2-trichloroacetic acid |
SMILES | C(=O)(C(Cl)(Cl)Cl)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10539770
|
Fisher Chemical
T/P500/50 |
250 g |
£62.05
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10549770
|
Fisher Chemical
T/P500/60 |
1 kg |
£159.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
76-03-9 | |
163.378 | |
YNJBWRMUSHSURL-UHFFFAOYSA-N | |
6421 | |
2,2,2-trichloroacetic acid |
C2HCl3O2 | |
4177 | |
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
CHEBI:30956 | |
C(=O)(C(Cl)(Cl)Cl)O |
Specifications
1.2 | |
52°C | |
White | |
4177 | |
YNJBWRMUSHSURL-UHFFFAOYSA-N | |
2,2,2-trichloroacetic acid | |
6421 | |
163.39g/mol | |
Solid |
76-03-9 | |
196°C | |
C2HCl3O2 | |
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
C(=O)(C(Cl)(Cl)Cl)O | |
163.378 | |
CHEBI:30956 | |
Plastic powder jar | |
1.2mbar at 50°C, 0.08mbar @25C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Trichloroacetic acid
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trichloroacetic Acid, for Electrophoresis, Fisher Chemical™