missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid Solution, MP Biomedicals™

Product Code. 11462691 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11462691

Brand: MP Biomedicals 0219605780

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Trichloroacetic acid Solution

  • It is used in protein precipitation; has been used to determine protein concentration by quantitative precipitation.
  • It is also Used as a decalcifier and fixative in microscopy.
  • A protocol for the precipitation of nucleic acids can be found in Molecular Cloning. Rats exposed to chronic sublethal amounts of trichloroacetic acid displayed an increase in serum bilirubin with a decrease in hematological proteins and cholesterols along with significant decreases in red blood corpuscles, mean cell volume, mean corpuscular hemoglobin, hemoglobin and hematocrit.
  • In mice the production of liver tumors was associated to Trichloroacetic acid, which is also a known mouse hepatocarinogen.
  • On the guinea pig trichloroacetic acid was observed to be a mild alergen on the skin.
  • Trichloroacetic acid is an acetic acid analogue commonly used to precipitate proteins, DNA and RNA. In the presence of SDS Trichloroacetic acid will precipitate proteins which can then be quantified by the Lowry method.
  • The compound has also found uses as a decalcifier and fixative in microscopy, in protein sequencing, detecting albumin and organic synthesis.
TRUSTED_SUSTAINABILITY

Specifications

pH 1.2 (0.1 M aqueous solution)(Literature)
CAS 76-03-9
Melting Point ∼56°C (Literature)
Molecular Formula C2HCl3O2
MDL Number MFCD00004177
UN Number UN2564
Beilstein 970119
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES C(=O)(C(Cl)(Cl)Cl)O
Concentration 100% w/v
IUPAC Name 2,2,2-trichloroacetic acid
Molecular Weight (g/mol) 163.4
PubChem CID 6421
ChEBI CHEBI:30956
Formula Weight 163.38
Density 1.62 sp. gr.
Quantity 100 mL
Physical Form Solution
Chemical Name or Material Trichloroacetic Acid Solution, 6.1N
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trichloroacetic Acid, 6.1 N Solution
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : trichloroacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.