missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetyl isocyanate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
Trichloroacetyl isocyanate is used as an in-situ derivatizing reagent for characterization of alcohols, glycols, phenols, and amines by nuclear magnetic resonance. It is also used in anion chemical ionization mass spectrometry and in catalytic one-pot dehydrative glycosylation of 1-hydroxy carbohydrate. Further, it serves as a reagent for the conversion of alcohols into carbamates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trichloroacetyl isocyanate is used as an in-situ derivatizing reagent for characterization of alcohols, glycols, phenols, and amines by nuclear magnetic resonance. It is also used in anion chemical ionization mass spectrometry and in catalytic one-pot dehydrative glycosylation of 1-hydroxy carbohydrate. Further, it serves as a reagent for the conversion of alcohols into carbamates.
Solubility
Miscible with dichloromethane, ether, terahydrofuran and protic solvents.
Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids and strong oxidizing agents.
Trichloroacetyl isocyanate is used as an in-situ derivatizing reagent for characterization of alcohols, glycols, phenols, and amines by nuclear magnetic resonance. It is also used in anion chemical ionization mass spectrometry and in catalytic one-pot dehydrative glycosylation of 1-hydroxy carbohydrate. Further, it serves as a reagent for the conversion of alcohols into carbamates.
Solubility
Miscible with dichloromethane, ether, terahydrofuran and protic solvents.
Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids and strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.585 |
Boiling Point | 58°C to 60°C (20 mmHg) |
Flash Point | 65°C (149°F) |
Odor | Acrid |
Refractive Index | 1.48 |
Quantity | 2 g |
UN Number | UN2927 |
Beilstein | 971201 |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible with dichloromethane,ether,terahydrofuran and protic solvents. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'