Learn More
Trichloroisocyanuric acid, 90+%, Thermo Scientific Chemicals
£15.80 - £147.00
Chemical Identifiers
CAS | 87-90-1 |
---|---|
Molecular Formula | C3Cl3N3O3 |
Molecular Weight (g/mol) | 232.40 |
MDL Number | MFCD00006553 |
InChI Key | YRIZYWQGELRKNT-UHFFFAOYSA-N |
Synonym | trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal |
PubChem CID | 6909 |
ChEBI | CHEBI:33015 |
SMILES | ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11406766
|
Thermo Scientific Alfa Aesar
B23906.22 |
100 g |
£15.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11416766
|
Thermo Scientific Alfa Aesar
B23906.36 |
500 g |
£45.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11426766
|
Thermo Scientific Alfa Aesar
B23906.0E |
2500 g |
£147.00
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Trichloroisocyanuric acid was used as a disinfection reagent in the transformation of benzophenone-3 in chlorinated water. It was also used in the synthesis of 2-halomethylene-3-oxoketoximes. It is used as a bleaching agent in the textile industry, a reagent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrichloroisocyanuric acid was used as a disinfection reagent in the transformation of benzophenone-3 in chlorinated water. It was also used in the synthesis of 2-halomethylene-3-oxoketoximes. It is used as a bleaching agent in the textile industry, a reagent in organic synthesis.
Solubility
Soluble in chlorocarbon, acetonitrile, acetone, water (12 g/L).
Notes
Moisture sensitive. Store away from reducing agents, acids, bases, flammable substances, organic materials, light, water/ moisture.
Chemical Identifiers
87-90-1 | |
232.40 | |
YRIZYWQGELRKNT-UHFFFAOYSA-N | |
6909 | |
ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O |
C3Cl3N3O3 | |
MFCD00006553 | |
trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal | |
CHEBI:33015 |
Specifications
87-90-1 | |
2.07 | |
>250°C (482°F) | |
C3Cl3N3O3 | |
UN2468 | |
Moisture sensitive | |
trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal | |
YRIZYWQGELRKNT-UHFFFAOYSA-N | |
trichloro-1,3,5-triazinane-2,4,6-trione | |
6909 | |
232.41 | |
Trichloroisocyanuric acid |
∼245°C (decomposition) | |
3 | |
Pungent | |
MFCD00006553 | |
202022 | |
14,9641 | |
Soluble in chlorocarbon,acetonitrile,acetone,water (12g/L). | |
ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O | |
232.40 | |
CHEBI:33015 | |
≥90% |
Safety and Handling
GHS H Statement
H272-H302-H319-H335
May intensify fire; oxidizer.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
H272-H302-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: TRICHLOROISOCYANURIC ACID, DRY
missing translation for 'einecsNumber' : 201-782-8
missing translation for 'rtecsNumber' : XZ1925000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only