missing translation for 'onlineSavingsMsg'
Learn More

Trichloromethyl chloroformate, 98%, Thermo Scientific Chemicals

£34.47 - £366.83

Chemical Identifiers

CAS 503-38-8
Molecular Formula C2Cl4O2
Molecular Weight (g/mol) 197.82
MDL Number MFCD00015553
InChI Key HCUYBXPSSCRKRF-UHFFFAOYSA-N
Synonym diphosgene, trichloromethyl chloroformate, diphosgen, difosgen czech, perchloromethyl formate, unii-po4q4r80lv, hsdb 371, carbonochloridic acid, trichloromethyl ester, methanol, trichloro-, chloroformate, chloroformic acid trichloromethyl ester
PubChem CID 10426
IUPAC Name trichloromethyl carbonochloridate
SMILES ClC(=O)OC(Cl)(Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11448037
SDS View Documents
Thermo Scientific Alfa Aesar
A17444.09
10 g
£34.47
10g
Please to purchase this item. Need a web account? Register with us today!
11458037
SDS View Documents
Thermo Scientific Alfa Aesar
A17444.18
50 g
£108.54
50g
Please to purchase this item. Need a web account? Register with us today!
11468037
SDS View Documents
Thermo Scientific Alfa Aesar
A17444.30
250 g
£366.83
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Trichloromethyl chloroformate is used as a reagent in the synthesis of organic compounds. It serves as a source of phosgene used in some laboratory preparations. Also, it is used as a reactant for the synthesis of cyclic carbamimidates, N-alkenyl and cycloalkyl carbamates and prostate-specific membrane antigen-targeted anticancer prodrugs. In addition, it is involved in the preparation of an erythromycin A derivatives and antibody-drug conjugates. It is utilized in the conversion of amines, carboxylic acids, formamides in to isocyanates, acid chlorides and isocyanides respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloromethyl chloroformate is used as a reagent in the synthesis of organic compounds. It serves as a source of phosgene used in some laboratory preparations. Also, it is used as a reactant for the synthesis of cyclic carbamimidates, N-alkenyl and cycloalkyl carbamates and prostate-specific membrane antigen-targeted anticancer prodrugs. In addition, it is involved in the preparation of an erythromycin A derivatives and antibody-drug conjugates. It is utilized in the conversion of amines, carboxylic acids, formamides in to isocyanates, acid chlorides and isocyanides respectively.

Solubility
Miscible with ethanol, ether and benzene. Immiscible with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.comStore in a cool place. Light and heat sensitive. Incompatible with strong oxidizing agents and bases.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H314-H318

P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300+H330-H314

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 207-965-9

missing translation for 'rtecsNumber' : LQ7350000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trichloromethyl chloroformate, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.