missing translation for 'onlineSavingsMsg'
Learn More

Tricine, 0.5M buffer soln., pH 6.5, Thermo Scientific™

£40.70 - £68.60

Chemical Identifiers

CAS 5704-04-1
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15462939
View Documents
Thermo Scientific Alfa Aesar
J60107.AE
100 mL
£40.70
100mL
Please to purchase this item. Need a web account? Register with us today!
15472939
View Documents
Thermo Scientific Alfa Aesar
J60107.AK
250 mL
£68.60
250mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5704-04-1
179.17
SEQKRHFRPICQDD-UHFFFAOYSA-N
CHEBI:39063
C(C(=O)O)NC(CO)(CO)CO
C6H13NO5
MFCD00004277
79784
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid

Specifications

5704-04-1
0.5 M
Liquid
MFCD00004277
C(C(=O)O)NC(CO)(CO)CO
179.17
CHEBI:39063
Colorless
6.5
C6H13NO5
SEQKRHFRPICQDD-UHFFFAOYSA-N
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
79784
Tricine buffer soln.
Videos
Safety and Handling

Safety and Handling

H335

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications