missing translation for 'onlineSavingsMsg'
Learn More

Tricine, 0.5M buffer soln., pH 7.0, Thermo Scientific™

Product Code. 15484639
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5704-04-1
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Color Colorless
Concentration 0.5 M
pH 7.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material Tricine buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'