missing translation for 'onlineSavingsMsg'
Learn More

Tricine, ≥98.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals

Product Code. 15835478
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 5704-04-1
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-Tris(Hydroxymethyl)methylglycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Melting Point ∼183°C (decomposition)
Physical Form Crystalline Powder
Quantity 500 g
Linear Formula (CH2OH)3CNHCH2COOH
Packaging Plastic bottle
Beilstein 1937804
Formula Weight 179.17
Percent Purity ≥98.5%
Assay Dry basis
Chemical Name or Material Tricine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'