missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tricine, 98%, for biochemistry

Product Code. 10498150
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5704-04-1
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine, N-[Tris(hydroxymethyl)methyl]glycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Absorbance 0.05 max. at 250nm
Absorbance Conditions (5% aq. soln.) (1 cm cell)
Melting Point 180.0°C to 184.0°C
Color White
pH 4.8 to 5.4 (0.5M aq. soln. at 20°C)
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 98%
Infrared Spectrum Authentic
Loss on Drying 1% max. (120°C)
Linear Formula (HOCH2)3CNHCH2CO2H
Packaging Plastic bottle
Merck Index 15, 9816
Solubility Information Solubility in water: soluble.
Formula Weight 179.17
Percent Purity 98%
Grade Biochemical
Chemical Name or Material Tricine, 98%
Show More Show Less
Product Identifier
  • Tricine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'