missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tricine, 99+%, for biochemistry
£30.45 - £1543.68
Chemical Identifiers
CAS | 5704-04-1 |
---|---|
Molecular Formula | C6H13NO5 |
Molecular Weight (g/mol) | 179.17 |
MDL Number | MFCD00004277 |
InChI Key | SEQKRHFRPICQDD-UHFFFAOYSA-N |
Synonym | N-[Tris(hydroxymethyl)methyl]glycine, N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine |
PubChem CID | 79784 |
ChEBI | CHEBI:39063 |
IUPAC Name | 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
SMILES | C(C(=O)O)NC(CO)(CO)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10027882
|
Thermo Scientific Chemicals
327830250 |
25 g | Plastic bottle |
£30.45
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10021811
|
Thermo Scientific Chemicals
327831000 |
100 g | Plastic bottle |
£85.43
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10742325
|
Thermo Scientific Chemicals
327830010 |
1 kg | Plastic bottle |
£502.50
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10676184
|
Thermo Scientific Chemicals
327830050 |
5 kg | Plastic drum |
£1543.68
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5704-04-1 | |
179.17 | |
SEQKRHFRPICQDD-UHFFFAOYSA-N | |
79784 | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
C6H13NO5 | |
MFCD00004277 | |
N-[Tris(hydroxymethyl)methyl]glycine, N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine | |
CHEBI:39063 | |
C(C(=O)O)NC(CO)(CO)CO |
Specifications
0.02 max. at 280nm | |
5704-04-1 | |
White | |
Crystalline Powder | |
Authentic | |
(HOCH2)3CNHCH2CO2H | |
N-[Tris(hydroxymethyl)methyl]glycine, N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine | |
SEQKRHFRPICQDD-UHFFFAOYSA-N | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid | |
79784 | |
179.17 | |
Biochemical |
(0.1M aq. soln.) | |
180.0°C to 184.0°C | |
4.6 to 5.6 (1% aq. soln. at 25°C) | |
99+% | |
C6H13NO5 | |
MFCD00004277 | |
Solubility in water: soluble. | |
C(C(=O)O)NC(CO)(CO)CO | |
179.17 | |
CHEBI:39063 | |
≥99% | |
Tricine |
Safety and Handling
Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation
Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Product Identifier
- Tricine
RUO – Research Use Only