missing translation for 'onlineSavingsMsg'
Learn More

Tricine, Electrophoresis Grade, MP Biomedicals

Catalog Number 11451632
Click to view available options
Quantity:
100 g
250 g
1 kg
This item is not returnable. View return policy
5704-04-1
C6H13NO5
179.17
SEQKRHFRPICQDD-UHFFFAOYSA-N
79784
CHEBI:39063
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C(C(=O)O)NC(CO)(CO)CO
This item is not returnable. View return policy
5704-04-1
C6H13NO5
179.17
SEQKRHFRPICQDD-UHFFFAOYSA-N
79784
CHEBI:39063
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C(C(=O)O)NC(CO)(CO)CO

5704-04-1 Purity: >>>99% Heavy metals (as Pb): <<10 ppm pKa at 25°C = 8.1 Biological buffer

Quantity 1 kg
Grade Electrophoresis
Chemical Name or Material Tricine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'