missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triclabendazole, 98%

Product Code. 15741789
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68786-66-3
Molecular Formula C14H9Cl3N2OS
Molecular Weight (g/mol) 359.66
InChI Key NQPDXQQQCQDHHW-UHFFFAOYSA-N
Synonym triclabendazole, fasinex, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzo d imidazole, triclabendazol inn-spanish, triclabendazolum inn-latin, ccris 8988, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-benzimidazole, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole
PubChem CID 50248
IUPAC Name 6-chloro-5-(2,3-dichlorophenoxy)-2-methylsulfanyl-1H-benzimidazole
SMILES CSC1=NC2=CC(=C(C=C2N1)Cl)OC3=C(C(=CC=C3)Cl)Cl

Specifications

Melting Point 175.0°C to 176.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 359.66
Percent Purity 98%
Chemical Name or Material Triclabendazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'