Learn More
Cytiva Tricorn™ Bottom Unit
Designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures
Brand:  Cytiva 18-1153-01
 View more versions of this product
Product Code. 10221030
  • £114.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures. They are suitable for use with high performance media, such as Sepharose High Performance, Superdex Prep Grade, and SOURCE.
  • The columns are available with inner diameters of 5 and 10mm, and in lengths from 20 to 200mm. Column lengths of 300 and 600mm are also available for the 10mm diameter columns. The design and dimensions of the columns make them particularly useful for screening and polishing steps in protein purification.
  • Glass tubes coated with plastic film protects individuals in the laboratory from harm if the column is dropped or operated outside its specification.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For 5 columns
Ethylene Propylene, PEEK, Polyethylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Tricorn™ Bottom Unit > For 5 columns