missing translation for 'onlineSavingsMsg'
Learn More
Tricyclohexylphosphine, 97%, Thermo Scientific Chemicals Product Code.: 10230041

Tricyclohexylphosphine, 97%, Thermo Scientific Chemicals

Product Code. 10230041
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10230041

Brand: Thermo Scientific Acros 421610050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2622-14-2
Molecular Formula C18H33P
Molecular Weight (g/mol) 280.42
MDL Number MFCD00003853
InChI Key WLPUWLXVBWGYMZ-UHFFFAOYSA-N
Synonym tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentrationShow MoreShow Less
PubChem CID 75806
IUPAC Name tricyclohexylphosphane
SMILES C1CCC(CC1)P(C2CCCCC2)C3CCCCC3

Specifications

Melting Point 77.0°C to 83.0°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 16, IV, 947
Merck Index 10, 9456
Formula Weight 280.42
Percent Purity 97%
Chemical Name or Material Tricyclohexylphosphine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0446115 Certificate TypeCertificate of Analysis Date25/04/2024 Catalog Number421610050,421610250,421610010
Lot NumberA0457176 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number421610050,421610250,421610010
Lot NumberA0457515 Certificate TypeCertificate of Analysis Date14/12/2023 Catalog Number421610050,421610250,421610010
Lot NumberA0397216 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number421610050,421610250,421610010
Lot NumberA0443489 Certificate TypeCertificate of Analysis Date13/09/2022 Catalog Number421610050,421610250,421610010
compliance-icons
Product Identifier
  • Tricyclohexyl phosphine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tricyclohexylphosphine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.