missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, 99+%, Thermo Scientific Chemicals

Product Code. 10793914
Click to view available options
Quantity:
25 L
Packaging:
Metal drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-71-6
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol
SMILES C(CO)N(CCO)CCO

Specifications

Melting Point 21.0°C
Color Colorless to Yellow
Density 1.1245g/mL
Boiling Point 360.0°C
Flash Point 190°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Metal drum
Molecular Formula C6H15NO3
Linear Formula (HOCH2CH2)3N
Refractive Index 1.4840 to 1.4860
Quantity 25 L
Fieser 01,1196
Merck Index 15, 9830
Specific Gravity 1.1245
Solubility Information Solubility in water: freely soluble. Other solubilities: miscible with alcohol,methanol and acetone,soluble in chloroform,slichtly soluble in benzene and ether,soluble 4.2% in benzene; 1.6% in ether; 0.4% in,carbon tetrachloride and < 0.1% in n-he
Viscosity 600 mPa.s (25°C)
Formula Weight 149.19
Percent Purity 99+%
Physical Form Oily Liquid
Chemical Name or Material Triethanolamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'