missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethanolamine solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£46.55 - £96.60
Chemical Identifiers
CAS | 9005-65-6 |
---|---|
Molecular Formula | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 |
Molecular Weight (g/mol) | 604.82 |
MDL Number | MFCD00082107 |
InChI Key | HDTIFOGXOGLRCB-KTKRTIGZNA-N |
IUPAC Name | 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |
SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12972215
|
Fisher Chemical
A 050 100 |
100 mL | Bottle |
£46.55
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12982215
|
Fisher Chemical
A 050 250 |
250 mL | Bottle |
£66.35
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12992215
|
Fisher Chemical
A 050 500 |
500 mL | Bottle |
£96.60
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
9005-65-6 | |
604.82 | |
HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
MFCD00082107 | |
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |
Specifications
1g/L | |
Single-component Standard Solution | |
MFCD00082107 | |
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
604.82 | |
Liquid |
9005-65-6 | |
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate | |
H2O | |
Triethanolamine |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)