missing translation for 'onlineSavingsMsg'
Learn More

Triethoxysilane, 95%, Thermo Scientific Chemicals

£37.08 - £377.88

Chemical Identifiers

CAS 998-30-1
Molecular Formula C6H16O3Si
Molecular Weight (g/mol) 164.28
MDL Number MFCD00009061
InChI Key PKDCQJMRWCHQOH-UHFFFAOYSA-N
Synonym triethoxysilane, silane, triethoxy, unii-8t460wdh89, c6h16o3si, si oet 3h;, c2h5o 3sih, 4-01-00-01359 beilstein handbook reference, triethoxysilane 10g, triethoxysilyl modified poly 1,2-butadiene, poly 1,2-butadiene , triethoxysilyl modified
PubChem CID 6327710
IUPAC Name triethoxysilicon
SMILES CCO[Si](OCC)OCC
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10163294
View Documents Promotion Details
Thermo Scientific Acros
164810100
10 g Glass bottle
£37.08
10g
Please to purchase this item. Need a web account? Register with us today!
10366834
View Documents
Thermo Scientific Acros
164810500
50 g Glass bottle
£116.58
50g
Please to purchase this item. Need a web account? Register with us today!
13428398
View Documents
Thermo Scientific Acros
164812500
250 g Glass bottle
£377.88
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

998-30-1
164.28
PKDCQJMRWCHQOH-UHFFFAOYSA-N
6327710
CCO[Si](OCC)OCC
C6H16O3Si
MFCD00009061
triethoxysilane, silane, triethoxy, unii-8t460wdh89, c6h16o3si, si oet 3h;, c2h5o 3sih, 4-01-00-01359 beilstein handbook reference, triethoxysilane 10g, triethoxysilyl modified poly 1,2-butadiene, poly 1,2-butadiene , triethoxysilyl modified
triethoxysilicon

Specifications

998-30-1
Colorless to Yellow
134.0°C to 135.0°C
Authentic
C6H16O3Si
1.3762
01,334
0.89
Solubility in water: reacts
CCO[Si](OCC)OCC
164.28
164.28
Liquid
-170.0°C
0.8900g/mL
26°C
94% min. (GC)
(C2H5O)3SiH
MFCD00009061
11,554; 17,366
triethoxysilane, silane, triethoxy, unii-8t460wdh89, c6h16o3si, si oet 3h;, c2h5o 3sih, 4-01-00-01359 beilstein handbook reference, triethoxysilane 10g, triethoxysilyl modified poly 1,2-butadiene, poly 1,2-butadiene , triethoxysilyl modified
PKDCQJMRWCHQOH-UHFFFAOYSA-N
triethoxysilicon
6327710
95%
Triethoxysilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 213-650-7

Product Identifier
  • Triethoxysilane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only