missing translation for 'onlineSavingsMsg'
Learn More
Triethyl borate, 97%, Thermo Scientific Chemicals Product Code.: 10031490

Triethyl borate, 97%, Thermo Scientific Chemicals

Product Code. 10031490
250 mL, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10031490

Brand: Thermo Scientific Acros 139582500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-46-9
Molecular Formula C6H15BO3
Molecular Weight (g/mol) 145.99
MDL Number MFCD00009073
InChI Key AJSTXXYNEIHPMD-UHFFFAOYSA-N
Synonym triethoxyborane, boron ethoxide, boron triethoxide, triethoxyboron, boric acid, triethyl ester, boric acid triethyl ester, borane, triethoxy, triethylborate, ethyl borate eto 3b, boric acid h3bo3 , triethyl ester
PubChem CID 9009
ChEBI CHEBI:38916
IUPAC Name triethyl borate
SMILES B(OCC)(OCC)OCC

Specifications

Melting Point -84.5°C
Color Colorless
Density 0.8580g/mL
Boiling Point 117.0°C to 118.0°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)3B
Refractive Index 1.3720 to 1.3740
Quantity 250 mL
Beilstein 01, 335
Specific Gravity 0.858
Solubility Information Solubility in water: decomposes
Formula Weight 145.99
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethyl borate, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465284 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog Number139582500,139580010
Lot NumberA0462061 Certificate TypeCertificate of Analysis Date22/04/2024 Catalog Number139582500,139580010
Lot NumberA0459228 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog Number139582500,139580010
Lot NumberA0457035 Certificate TypeCertificate of Analysis Date19/10/2023 Catalog Number139582500,139580010
Lot NumberA0456125 Certificate TypeCertificate of Analysis Date06/09/2023 Catalog Number139582500,139580010
compliance-icons
Product Identifier
  • Triethyl borate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triethyl borate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.