missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl borate, 97%
Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Förpackningsstorlek
1 L
250mL
Beskrivning
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Melting Point | -84.5°C |
| Color | Colorless |
| Density | 0.8580g/mL |
| Boiling Point | 117.0°C to 118.0°C |
| Flash Point | 11°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | (C2H5O)3B |
| Refractive Index | 1.3720 to 1.3740 |
| Visa mer |
Säkerhet och hantering
Produktbeteckning
- Triethyl borate
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering