missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 77-93-0 |
---|---|
Molecular Formula | C12H20O7 |
Molecular Weight (g/mol) | 276.29 |
InChI Key | DOOTYTYQINUNNV-UHFFFAOYSA-N |
Synonym | triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester |
PubChem CID | 6506 |
IUPAC Name | triethyl 2-hydroxypropane-1,2,3-tricarboxylate |
SMILES | CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10075942
|
Thermo Scientific Acros
375062500 |
250 g | Plastic bottle |
£22.91
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10137742
|
Thermo Scientific Acros
375060010 |
1 kg | Plastic bottle |
£73.57
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11995061
|
Thermo Scientific Acros
375060025 |
2.5 kg | Plastic bottle |
£152.76
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
77-93-0 | |
276.29 | |
triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester | |
triethyl 2-hydroxypropane-1,2,3-tricarboxylate |
C12H20O7 | |
DOOTYTYQINUNNV-UHFFFAOYSA-N | |
6506 | |
CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O |
Specifications
77-93-0 | |
1.1360g/mL | |
151°C | |
98.5% min. (GC) | |
1.4400 to 1.4430 | |
1.136 | |
Solubility in water: 5.7% (25°C) | |
CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O | |
276.29 | |
35.2 mPa.s (25°C) | |
99% | |
Triethyl citrate |
-46.0°C | |
294.0°C | |
Authentic | |
C12H20O7 | |
15,2325 | |
triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester | |
DOOTYTYQINUNNV-UHFFFAOYSA-N | |
triethyl 2-hydroxypropane-1,2,3-tricarboxylate | |
6506 | |
276.29 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 201-070-7
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title