missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl orthoformate, 98%, Thermo Scientific Chemicals
£16.20 - £673.00
Chemical Identifiers
CAS | 122-51-0 |
---|---|
Molecular Formula | C7H16O3 |
Molecular Weight (g/mol) | 148.2 |
MDL Number | MFCD00009230 |
InChI Key | GKASDNZWUGIAMG-UHFFFAOYSA-N |
Synonym | triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty |
PubChem CID | 31214 |
IUPAC Name | diethoxymethoxyethane |
SMILES | CCOC(OCC)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10512541
|
Thermo Scientific Acros
139661000 |
100 mL | Glass bottle |
£16.20
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10476960
|
Thermo Scientific Acros
139660010 |
1 L | Glass bottle |
£51.50
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10225430
|
Thermo Scientific Acros
139660025 |
2.5 L | Glass bottle |
£100.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10297133
|
Thermo Scientific Acros
139660250 |
25 L | Metal Drum |
£673.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
122-51-0 | |
148.2 | |
GKASDNZWUGIAMG-UHFFFAOYSA-N | |
31214 | |
CCOC(OCC)OCC |
C7H16O3 | |
MFCD00009230 | |
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty | |
diethoxymethoxyethane |
Specifications
122-51-0 | |
Colorless | |
146.0°C | |
Authentic | |
C7H16O3 | |
1.3900 to 1.3920 | |
02, 20 | |
15, 6980 | |
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty | |
GKASDNZWUGIAMG-UHFFFAOYSA-N | |
diethoxymethoxyethane | |
31214 | |
98% | |
Triethyl orthoformate |
-61.0°C | |
0.8900g/mL | |
35°C | |
97.5% min. (GC) | |
CH(OC2H5)3 | |
MFCD00009230 | |
01,1204; 02,430; 04,527; 05,690; 06,610; 07,385; 11,555; 13,327 | |
0.89 | |
Solubility in water: 1.35g/L (20°C). Other solubilities: decomposes in water,soluble in alcohols,acetone,ethers,chloroform, | |
CCOC(OCC)OCC | |
148.2 | |
148.2 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-550-4
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Triethyl orthoformate
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only