missing translation for 'onlineSavingsMsg'
Learn More
Triethyl phosphate, 99%, Thermo Scientific Chemicals Product Code.: 10522731

Triethyl phosphate, 99%, Thermo Scientific Chemicals

Product Code. 10522731
1 kg, Glass bottle
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10522731

Brand: Thermo Scientific Acros 139680010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-40-0
Molecular Formula C6H15O4P
Molecular Weight (g/mol) 182.16
MDL Number MFCD00009077
InChI Key DQWPFSLDHJDLRL-UHFFFAOYSA-N
Synonym triethylphosphate, phosphoric acid, triethyl ester, tris ethyl phosphate, triethoxyphosphine oxide, triethylfosfat, ethyl phosphate eto 3po, triethylfosfat czech, ethyl phosphate van, unii-qih4k96k7j, phosphoric acid triethyl ester
PubChem CID 6535
ChEBI CHEBI:45927
IUPAC Name triethyl phosphate
SMILES CCOP(=O)(OCC)OCC

Specifications

Melting Point -56°C
Density 1.0700g/mL
Boiling Point 215°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)3P(O)
Refractive Index 1.4050 to 1.407
Quantity 1 kg
Beilstein 01, 332
Fieser 01,1212; 16,3
Merck Index 15, 9838
Specific Gravity 1.07
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols,ethers,ketones,esters,,aliphatic,aromatic and chlorinated hydrocarbons,soluble in most other organic solvents
Formula Weight 182.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triethyl phosphate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0403317 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number139680010,139680025
Lot NumberA0412226 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number
Lot NumberA0442865 Certificate TypeCertificate of Analysis Date06/06/2022 Catalog Number
Lot NumberA0434002 Certificate TypeCertificate of Analysis Date14/09/2021 Catalog NumberC22322.30
Lot NumberA0430165 Certificate TypeCertificate of Analysis Date18/05/2021 Catalog NumberC22322.30
compliance-icons
Product Identifier
  • Triethyl phosphate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triethyl phosphate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.