missing translation for 'onlineSavingsMsg'
Learn More

Triethylaluminium, 1.3M solution in Heptane, Thermo Scientific Chemicals

Product Code. 10172172
Click to view available options
Quantity:
100 mL
800 mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-93-8, 142-82-5
Molecular Formula C6H15Al
MDL Number MFCD00009015
InChI Key VOITXYVAKOUIBA-UHFFFAOYSA-N
Synonym triethylaluminum, triethylaluminium, aluminum, triethyl, triethylalane, unii-h426e9h3tt, triethylaluminum solution, 1.0 m in hexanes, triethylaluminum solution, 25 wt. % in toluene, triethyl-aluminum, triethyl aluminum, triethyl-aluminum
PubChem CID 16682930
IUPAC Name triethylalumane
SMILES CC[Al](CC)CC

Specifications

Color Colorless
Linear Formula (C2H5)3Al
Concentration 19.5 to 20.5 wt% (on Al basis)
Density 0.7300g/mL
Molecular Weight (g/mol) 114.168
Formula Weight 114.17
Packaging AcroSeal glass bottle
Quantity 100 mL
Physical Form Solution
Chemical Name or Material Triethylaluminium, 1.3M Solution in Heptane
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triethylaluminium, 1.M solution in heptane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.