missing translation for 'onlineSavingsMsg'
Learn More
Triethylamine, 99.7%, extra pure, Thermo Scientific Chemicals Product Code.: 10741301

Triethylamine, 99.7%, extra pure, Thermo Scientific Chemicals

Product Code. 10741301
50 g
Click to view available options
Quantity:
5 g
50 g
1 kg
2.5 kg
10 kg
This item is not returnable. View return policy

Product Code. 10741301

Brand: Thermo Scientific Acros 219510500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8
MDL Number MFCD00009051
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Melting Point -115.0°C
Density 0.7280g/mL
Boiling Point 90.0°C
Flash Point −11°C
Infrared Spectrum Authentic
Assay Percent Range 99.7%
Molecular Formula C6H15N
Linear Formula (C2H5)3N
Refractive Index 1.3990 to 1.4010
Quantity 50 g
Beilstein 04, 99
Fieser 01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336
Merck Index 15, 9831
Specific Gravity 0.728
Solubility Information Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride
Molecular Weight (g/mol) 101.193
Formula Weight 101.19
Vapor Pressure 69mbar at 20°C
Percent Purity 99.7%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Triethylamine, 99.7%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0436271 Certificate TypeCertificate of Analysis Date11/11/2024 Catalog Number219510010,219510100,219510050,219510025,219510500
Lot NumberA0434813 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog Number219510010,219510100,219510050,219510025,219510500
Lot NumberB0546681B Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number219510010,219510100,219510050,219510025,219510500
Lot NumberA0463505 Certificate TypeCertificate of Analysis Date04/06/2024 Catalog Number219510010,219510100,219510050,219510025,219510500
Lot NumberB0541995A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number219510010,219510100,219510050,219510025,219510500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triethylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triethylamine, 99.7%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.