Learn More
Triethylamine, 99%, Thermo Scientific Chemicals
£24.64 - £65.32
Chemical Identifiers
CAS | 121-44-8 |
---|---|
Molecular Formula | C6H15N |
Molecular Weight (g/mol) | 101.193 |
MDL Number | MFCD00009051 |
InChI Key | ZMANZCXQSJIPKH-UHFFFAOYSA-N |
Synonym | triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethaneShow More |
PubChem CID | 8471 |
ChEBI | CHEBI:35026 |
IUPAC Name | N,N-diethylethanamine |
SMILES | CCN(CC)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413920
|
Thermo Scientific Alfa Aesar
A12646.AK |
250 mL |
£24.64
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470493
|
Thermo Scientific Alfa Aesar
A12646.AP |
500 mL |
£35.68
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480493
|
Thermo Scientific Alfa Aesar
A12646.0F |
2500 mL |
£65.32
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizer for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in automotive casting industry and textile industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizer for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in automotive casting industry and textile industry.
Solubility
Miscible with water, ether and ethanol.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
121-44-8 | |
101.193 | |
ZMANZCXQSJIPKH-UHFFFAOYSA-N | |
8471 | |
N,N-diethylethanamine |
C6H15N | |
MFCD00009051 | |
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethaneShow More | |
CHEBI:35026 | |
CCN(CC)CC |
Specifications
121-44-8 | |
0.726 | |
−11°C (12°F) | |
Ammonia-like | |
(CH3CH2)3N | |
MFCD00009051 | |
1843166 | |
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane | |
ZMANZCXQSJIPKH-UHFFFAOYSA-N | |
N,N-diethylethanamine | |
8471 | |
101.19 | |
Liquid |
-115°C | |
89°C to 90°C | |
99% | |
C6H15N | |
1.4005 | |
UN1296 | |
14,9666 | |
Miscible with water,ether and ethanol. | |
CCN(CC)CC | |
101.193 | |
CHEBI:35026 | |
99% | |
Triethylamine |
Safety and Handling
GHS H Statement
H225-H314-H302-H312-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H311+H331-H314-H335-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIETHYLAMINE
missing translation for 'einecsNumber' : 204-469-4
missing translation for 'rtecsNumber' : YE0175000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Triethylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : triethylamine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.