missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, 99%, pure, Thermo Scientific Chemicals

Product Code. 10651193
Click to view available options
Quantity:
5 mL
100 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Melting Point -115.0°C
Density 0.7280g/mL
Boiling Point 90.0°C
Flash Point −11°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H15N
Linear Formula (C2H5)3N
Refractive Index 1.3990 to 1.4010
Quantity 100 mL
Beilstein 04, 99
Fieser 01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336
Merck Index 15, 9831
Specific Gravity 0.728
Solubility Information Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride
Molecular Weight (g/mol) 101.193
Formula Weight 101.19
Vapor Pressure 69mbar at 20°C
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Triethylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triethylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'