missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylamine, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Description
Triethylamine, Certified AR for Analysis
Specifications
Specifications
Melting Point | -115°C |
Color | Colorless |
pH | 12.4 |
Boiling Point | 90°C |
Packaging | Plastic Coated Glass bottle |
Quantity | 2.5 L |
Vapor Density | 3.5 |
Formula Weight | 101.19g/mol |
Vapor Pressure | 69mbar at 20°C |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Triethylamine
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'