Learn More
Triethylamine trihydrofluoride, ca. 37% HF, Thermo Scientific Chemicals
£62.00 - £160.00
Chemical Identifiers
CAS | 73602-61-6 |
---|---|
Molecular Formula | C6H15N·3HF |
Molecular Weight (g/mol) | 161.21 |
MDL Number | MFCD00043294 |
InChI Key | IKGLACJFEHSFNN-UHFFFAOYSA-N |
Synonym | triethylamine trihydrofluoride, n,n-diethylethanamine trihydrofluoride, triethylammonium fluoride, ethanamine, n,n-diethyl-, trihydrofluoride, triethylamine trishydrofluoride, hydrogen fluoride in triethylamine, hf in triethylamine, triethylaminetrihydrofluoride, hydrogen fluoride triethylamine, ethanamine, n,n-diethyl-, hydrofluoride 1:3 |
PubChem CID | 175505 |
IUPAC Name | N,N-diethylethanamine;trihydrofluoride |
SMILES | CCN(CC)CC.F.F.F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10081601
|
Thermo Scientific Acros
350620250 |
25 mL | Plastic bottle |
£62.00
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10511194
|
Thermo Scientific Acros
350621000 |
100 mL | Plastic bottle |
£160.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentIdentifiants chimiques
73602-61-6 | |
161.21 | |
IKGLACJFEHSFNN-UHFFFAOYSA-N | |
175505 | |
CCN(CC)CC.F.F.F |
C6H15N·3HF | |
MFCD00043294 | |
triethylamine trihydrofluoride, n,n-diethylethanamine trihydrofluoride, triethylammonium fluoride, ethanamine, n,n-diethyl-, trihydrofluoride, triethylamine trishydrofluoride, hydrogen fluoride in triethylamine, hf in triethylamine, triethylaminetrihydrofluoride, hydrogen fluoride triethylamine, ethanamine, n,n-diethyl-, hydrofluoride 1:3 | |
N,N-diethylethanamine;trihydrofluoride |
Spécification
73602-61-6 | |
Yellow to Brown | |
70.0°C (15.0 mmHg) | |
Authentic | |
C6H15N·3HF | |
1.3860 to 1.3920 | |
0.989 | |
Solubility in water: soluble | |
CCN(CC)CC.F.F.F | |
161.21 | |
161.21 | |
Liquid |
-29.0°C | |
0.9890g/mL | |
87°C | |
97% | |
(C2H5)3N·3HF | |
MFCD00043294 | |
triethylamine trihydrofluoride, n,n-diethylethanamine trihydrofluoride, triethylammonium fluoride, ethanamine, n,n-diethyl-, trihydrofluoride, triethylamine trishydrofluoride, hydrogen fluoride in triethylamine, hf in triethylamine, triethylaminetrihydrofluoride, hydrogen fluoride triethylamine, ethanamine, n,n-diethyl-, hydrofluoride 1:3 | |
IKGLACJFEHSFNN-UHFFFAOYSA-N | |
N,N-diethylethanamine;trihydrofluoride | |
175505 | |
97% | |
Triethylamine trihydrofluoride, ca. 37% HF |
Sécurité et traitement
GHS H Statement
Causes severe skin burns and eye damage.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 277-550-5
- Triethylamine trihydrofluoride
Mention d'avertissement
- Danger
Catégorie de danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Mention de danger
- H310-Fatal in contact with skin.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H300-Fatal if swallowed.
Conseils de prudence
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only