missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Fluka™ Triethylammonium Phosphate Solution, pH 3, Honeywell™ Fluka™
BioUltra, ∼1.0 M in H2O
£80.14 - £312.87
Specifications
Molecular Weight (g/mol) | 199.187 |
---|---|
pH | 2.9 to 3 |
Packaging | Glass bottle |
CAS | 10138-93-9 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15657420
|
Honeywell Fluka™
90361-100ML |
100 mL | Glass bottle |
£80.14
100mL |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15667420
|
Honeywell Fluka™
90361-500ML |
500 mL | Glass bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Finest quality products for life science
- The most stringent testing protocols and highest purity
- Superior manufacturing methods and product specific testing protocols designed to ensure quality to our highest standard
- Tests may include, but are not limited to, purity by gel electrophoresis, trace metal testing and application testing.
Specifications
199.187 | |
2.9 to 3 | |
Glass bottle | |
10138-93-9 |