missing translation for 'onlineSavingsMsg'
Learn More

Triethylene glycol, 99%

Product Code. 10617934 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
This item is not returnable. View return policy

Product Code. 10617934

Brand: Thermo Scientific Acros 139590100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-27-6 | C6H14O4 | 150.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-27-6
Molecular Formula C6H14O4
Molecular Weight (g/mol) 150.17
MDL Number MFCD00002880,MFCD00081839,MFCD01779596,MFCD01779599,MFCD01779601,MFCD01779603,MFCD01779605,MFCD01779609,MFCD01779611,MFCD01779612,MFCD01779614,MFCD01779615,MFCD01779616
InChI Key ZIBGPFATKBEMQZ-UHFFFAOYSA-N
Synonym triethylene glycol, triglycol, 2,2'-ethane-1,2-diylbis oxy diethanol, trigen, triethylenglykol, triethyleneglycol, 2,2'-ethylenedioxydiethanol, 1,2-bis 2-hydroxyethoxy ethane, 3,6-dioxaoctane-1,8-diol, 2,2'-ethylenedioxy diethanol
PubChem CID 8172
ChEBI CHEBI:44926
SMILES OCCOCCOCCO

Specifications

Melting Point -7.0°C
Color Yellow
Density 1.1200g/mL
Boiling Point 285.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Linear Formula HOCH2CH2OCH2CH2OCH2CH2OH
Refractive Index 1.4545 to 1.4565
Quantity 10 L
Beilstein 01, IV, 2400
Merck Index 15, 9835
Specific Gravity 1.12
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol,benzene and toluene,sparingly soluble in ether,insoluble in petroleum ether,aliphatic,hydrocarbons
IUPAC Name 2-[2-(2-hydroxyethoxy)ethoxy]ethan-1-ol
Viscosity 48 mPa.s (20°C)
Formula Weight 150.17
Vapor Pressure <0.01mbar at 20°C
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material Triethylene glycol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.