missing translation for 'onlineSavingsMsg'
Learn More

Triethylenetetramine, 60%, Thermo Scientific Chemicals

Product Code. 10633522
Click to view available options
Quantity:
50 g
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-24-3
Molecular Formula C6H18N4
Molecular Weight (g/mol) 146.24
MDL Number MFCD00008169
InChI Key VILCJCGEZXAXTO-UHFFFAOYSA-N
Synonym triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
PubChem CID 5565
ChEBI CHEBI:39501
IUPAC Name N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
SMILES C(CNCCNCCN)N

Specifications

Melting Point -35.0°C
Density 0.9800g/mL
Boiling Point 280.0°C
Flash Point 129°C
Infrared Spectrum Authentic
Assay Percent Range 55% min. (GC)
Packaging Glass bottle
Linear Formula H2NCH2CH2NHCH2CH2NHCH2CH2NH2
Refractive Index 1.4960 to 1.5000
Quantity 50 g
Beilstein 04,255
Fieser 01,1204
Merck Index 15,9828
Specific Gravity 0.98
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol
Formula Weight 146.24
Percent Purity 60%
Grade Technical
Physical Form Liquid
Chemical Name or Material Triethylenetetramine, 60%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triethylenetetramine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'