Learn More
Triethylenetetramine, tech. 60%, balance branched and cyclic triethylenetetramines, Thermo Scientific Chemicals
£36.18 - £116.58
Chemical Identifiers
CAS | 112-24-3 |
---|---|
Molecular Formula | C6H18N4 |
Molecular Weight (g/mol) | 146.238 |
MDL Number | MFCD00008169 |
InChI Key | VILCJCGEZXAXTO-UHFFFAOYSA-N |
Synonym | triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951 |
PubChem CID | 5565 |
ChEBI | CHEBI:39501 |
IUPAC Name | N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine |
SMILES | C(CNCCNCCN)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11463167
|
Thermo Scientific Alfa Aesar
A16128.AP |
500 mL |
£36.18
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473167
|
Thermo Scientific Alfa Aesar
A16128.0F |
2500 mL |
£116.58
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Catalyzes the hydrolysis of N-terminal peptide bonds. Determination of the cation exchange capacity (CEC) of clay minerals is by using the complexes of copper(II) ion with triethylenetetramine and tetraethylenepentamine. Variable spin iron(iii) chelates with hexadentate ligands are derived from triethylenetetramine and various salicylaldehydes. Adsorption of Cu(II), Cd(II) and Pb(II) from aqueous single metal solutions by succinylated twice-mercerized sugarcane bagasse functionalized with triethylenetetramine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCatalyzes the hydrolysis of N-terminal peptide bonds. Determination of the cation exchange capacity (CEC) of clay minerals is by using the complexes of copper(II) ion with triethylenetetramine and tetraethylenepentamine. Variable spin iron(iii) chelates with hexadentate ligands are derived from triethylenetetramine and various salicylaldehydes. Adsorption of Cu(II), Cd(II) and Pb(II) from aqueous single metal solutions by succinylated twice-mercerized sugarcane bagasse functionalized with triethylenetetramine.
Solubility
Insoluble in water.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Chemical Identifiers
112-24-3 | |
146.238 | |
VILCJCGEZXAXTO-UHFFFAOYSA-N | |
5565 | |
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine |
C6H18N4 | |
MFCD00008169 | |
triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951 | |
CHEBI:39501 | |
C(CNCCNCCN)N |
Specifications
112-24-3 | |
0.982 | |
122°C (252°F) | |
C6H18N4 | |
1.498 | |
UN2259 | |
Hygroscopic | |
triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951 | |
VILCJCGEZXAXTO-UHFFFAOYSA-N | |
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine | |
5565 | |
146.24 | |
Technical | |
Triethylenetetramine |
∼-35°C | |
∼267°C | |
Amine-like | |
H2NCH2CH2(NHCH2CH2)2NH2 | |
MFCD00008169 | |
605448 | |
14,9663 | |
Insoluble in water. | |
C(CNCCNCCN)N | |
146.238 | |
CHEBI:39501 | |
60% | |
balance branched and cyclic triethylenetetramines |
Safety and Handling
GHS H Statement
H314-H312-H317
Causes severe skin burns and eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.
P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302+H332-H311-H314-H317-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TRIETHYLENETETRAMINE
missing translation for 'einecsNumber' : 203-950-6
missing translation for 'rtecsNumber' : YE6650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H371-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only