Learn More
Triethylgermanium chloride, Thermo Scientific™
£42.90 - £131.00
Chemical Identifiers
CAS | 994-28-5 |
---|---|
Molecular Formula | C6H17ClGe |
Molecular Weight (g/mol) | 197.28 |
MDL Number | MFCD00013591 |
InChI Key | FJAWDWVPBHGANJ-UHFFFAOYSA-N |
Synonym | triethylgermanium chloride, triethylchlorogermane, chloro triethyl germane, triethylchlorogermene, ccris 2650, triethylgermyl chloride, germanium triethyl chloride, acmc-209sco, chlorotriethylgermane |
PubChem CID | 70438 |
IUPAC Name | chloro(triethyl)germane |
SMILES | Cl.CC[GeH](CC)CC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
994-28-5 | |
197.28 | |
FJAWDWVPBHGANJ-UHFFFAOYSA-N | |
70438 | |
Cl.CC[GeH](CC)CC |
C6H17ClGe | |
MFCD00013591 | |
triethylgermanium chloride, triethylchlorogermane, chloro triethyl germane, triethylchlorogermene, ccris 2650, triethylgermyl chloride, germanium triethyl chloride, acmc-209sco, chlorotriethylgermane | |
chloro(triethyl)germane |
Specifications
994-28-5 | |
1.18 g/mL | |
C6H17ClGe | |
UN3265 | |
triethylgermanium chloride, triethylchlorogermane, chloro triethyl germane, triethylchlorogermene, ccris 2650, triethylgermyl chloride, germanium triethyl chloride, acmc-209sco, chlorotriethylgermane | |
FJAWDWVPBHGANJ-UHFFFAOYSA-N | |
chloro(triethyl)germane | |
70438 | |
Liquid |
<-50°C | |
175.9°C | |
MFCD00013591 | |
Moisture sensitive | |
Insoluble in water. | |
Cl.CC[GeH](CC)CC | |
197.28 | |
195.23 | |
Triethylgermanium chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 213-614-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures