missing translation for 'onlineSavingsMsg'
Learn More

Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride, Thermo Scientific Chemicals

£34.27 - £253.33

Specifications

Chemical Name or Material Triethyloxonium tetrafluoroborate
Name Note 1M Solution in Methylene Chloride
CAS 368-39-8, 75-09-2
Molecular Formula C6H15BF4O
Linear Formula (C2H5)3OBF4
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10695422
View Documents Promotion Details
Thermo Scientific Acros
175110250
25 mL
£34.27
25mL
Please to purchase this item. Need a web account? Register with us today!
10133860
View Documents Promotion Details
Thermo Scientific Acros
175111000
100 mL
£87.03
100mL
Please to purchase this item. Need a web account? Register with us today!
11905811
View Documents Promotion Details
Thermo Scientific Acros
175115000
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

Triethyloxonium tetrafluoroborate
368-39-8, 75-09-2
(C2H5)3OBF4
01,1210; 02,430; 03,303; 04,527; 05,691; 06,611; 07,386; 08,500; 09,482; 10,417; 15,337
triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
tetrafluoroboranuide; triethyloxidanium
2723982
1.328
1M Solution in Methylene Chloride
C6H15BF4O
MFCD00044423
1.3280g/mL
Solubility in water: reacts.
F[B-](F)(F)F.CC[O+](CC)CC
189.99
189.99
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing cancer.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'einecsNumber' : 206-705-1

missing translation for 'tsca' : TSCA

Product Identifier
  • Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only