Learn More
Triethylsilanol, 97%, Thermo Scientific Chemicals
£31.36 - £88.84
Chemical Identifiers
CAS | 597-52-4 |
---|---|
Molecular Formula | C6H16OSi |
Molecular Weight (g/mol) | 132.278 |
MDL Number | MFCD00042640 |
InChI Key | WVMSIBFANXCZKT-UHFFFAOYSA-N |
Synonym | triethylsilanol, silanol, triethyl, hydroxytriethylsilane, silanol, 1,1,1-triethyl, triethyl hydroxy silane, triethyl hydroxy silicon, tesoh, c2h5 3sioh, acmc-1an47 |
PubChem CID | 69005 |
IUPAC Name | triethyl(hydroxy)silane |
SMILES | CC[Si](CC)(CC)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322286
|
Thermo Scientific Alfa Aesar
L03738.03 |
1 g |
£31.36
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332286
|
Thermo Scientific Alfa Aesar
L03738.06 |
5 g |
£88.84
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
597-52-4 | |
132.278 | |
WVMSIBFANXCZKT-UHFFFAOYSA-N | |
69005 | |
CC[Si](CC)(CC)O |
C6H16OSi | |
MFCD00042640 | |
triethylsilanol, silanol, triethyl, hydroxytriethylsilane, silanol, 1,1,1-triethyl, triethyl hydroxy silane, triethyl hydroxy silicon, tesoh, c2h5 3sioh, acmc-1an47 | |
triethyl(hydroxy)silane |
Specifications
154°C | |
C6H16OSi | |
UN1993 | |
57°C (135°F) | |
WVMSIBFANXCZKT-UHFFFAOYSA-N | |
triethyl(hydroxy)silane | |
69005 | |
97% | |
1.433 | |
Triethylsilanol |
597-52-4 | |
MFCD00042640 | |
1732848 | |
triethylsilanol, silanol, triethyl, hydroxytriethylsilane, silanol, 1,1,1-triethyl, triethyl hydroxy silane, triethyl hydroxy silicon, tesoh, c2h5 3sioh, acmc-1an47 | |
CC[Si](CC)(CC)O | |
132.278 | |
132.28 | |
Mild | |
0.86 |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 209-903-6
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
- Triethylsilanol
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : Triethylsilanol
RUO – Research Use Only