Learn More
Triethylsilyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals
£41.50 - £138.00
Chemical Identifiers
CAS | 79271-56-0 |
---|---|
Molecular Formula | C7H15F3O3SSi |
Molecular Weight (g/mol) | 264.33 |
MDL Number | MFCD00000407 |
InChI Key | STMPXDBGVJZCEX-UHFFFAOYSA-N |
Synonym | triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf |
PubChem CID | 2733308 |
IUPAC Name | triethylsilyl trifluoromethanesulfonate |
SMILES | CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10033650
|
Thermo Scientific Acros
312700100 |
10 g | Glass bottle |
£41.50
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11975831
|
Thermo Scientific Acros
312700500 |
50 g | Glass bottle |
£138.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagentChemical Identifiers
79271-56-0 | |
264.33 | |
STMPXDBGVJZCEX-UHFFFAOYSA-N | |
2733308 | |
CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F |
C7H15F3O3SSi | |
MFCD00000407 | |
triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf | |
triethylsilyl trifluoromethanesulfonate |
Specifications
79271-56-0 | |
1.1600g/mL | |
72°C | |
99% | |
CF3SO3Si(C2H5)3 | |
MFCD00000407 | |
triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf | |
STMPXDBGVJZCEX-UHFFFAOYSA-N | |
triethylsilyl trifluoromethanesulfonate | |
2733308 | |
99% | |
Triethylsilyl trifluoromethanesulfonate |
Colorless to Brown | |
85.0°C to 86.0°C (12.0 mmHg) | |
Authentic | |
C7H15F3O3SSi | |
1.3890 to 1.3920 | |
1.16 | |
Solubility in water: soluble | |
CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F | |
264.33 | |
264.34 | |
Fuming Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 279-124-4
RUO – Research Use Only