Learn More
(α,α,α-Trifluoro-p-tolyl)acetic acid, 98%, Thermo Scientific Chemicals
£24.80 - £72.80
Chemical Identifiers
CAS | 32857-62-8 |
---|---|
Molecular Formula | C9H7F3O2 |
Molecular Weight (g/mol) | 204.15 |
MDL Number | MFCD00004352 |
InChI Key | HNORVZDAANCHAY-UHFFFAOYSA-N |
Synonym | 4-trifluoromethyl phenylacetic acid, 2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylphenylacetic acid, 4-trifluoromethyl phenyl acetic acid, alpha,alpha,alpha-trifluoro-p-tolyl acetic acid, benzeneacetic acid, 4-trifluoromethyl, p-trifluoromethyl phenylacetic acid, p-trifluoromethylphenylacetic acid, alpha,alpha,alpha-trifluoro-4-tolyl acetic acid |
PubChem CID | 118342 |
IUPAC Name | 2-[4-(trifluoromethyl)phenyl]acetic acid |
SMILES | OC(=O)CC1=CC=C(C=C1)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10647182
|
Thermo Scientific Acros
214370010 |
1 g | Glass bottle |
£24.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10361514
|
Thermo Scientific Acros
214370050 |
5 g | Glass bottle |
£72.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
32857-62-8 | |
204.15 | |
HNORVZDAANCHAY-UHFFFAOYSA-N | |
118342 | |
OC(=O)CC1=CC=C(C=C1)C(F)(F)F |
C9H7F3O2 | |
MFCD00004352 | |
4-trifluoromethyl phenylacetic acid, 2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylphenylacetic acid, 4-trifluoromethyl phenyl acetic acid, alpha,alpha,alpha-trifluoro-p-tolyl acetic acid, benzeneacetic acid, 4-trifluoromethyl, p-trifluoromethyl phenylacetic acid, p-trifluoromethylphenylacetic acid, alpha,alpha,alpha-trifluoro-4-tolyl acetic acid | |
2-[4-(trifluoromethyl)phenyl]acetic acid |
Specifications
32857-62-8 | |
100.0 | |
White to Yellow | |
98% | |
CF3C6H4CH2CO2H | |
4-trifluoromethyl phenylacetic acid, 2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylphenylacetic acid, 4-trifluoromethyl phenyl acetic acid, alpha,alpha,alpha-trifluoro-p-tolyl acetic acid, benzeneacetic acid, 4-trifluoromethyl, p-trifluoromethyl phenylacetic acid, p-trifluoromethylphenylacetic acid, alpha,alpha,alpha-trifluoro-4-tolyl acetic acid | |
OC(=O)CC1=CC=C(C=C1)C(F)(F)F | |
204.15 | |
204.15 | |
Crystalline Powder or Crystals |
97.5 | |
81°C to 85°C | |
Authentic | |
C9H7F3O2 | |
MFCD00004352 | |
HNORVZDAANCHAY-UHFFFAOYSA-N | |
2-[4-(trifluoromethyl)phenyl]acetic acid | |
118342 | |
98% | |
(α, α, α-Trifluoro-p-tolyl)acetic acid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 251-263-5
RUO – Research Use Only