missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid-d, 99.5%(Isotopic), Thermo Scientific Chemicals

Product Code. 11324468
Click to view available options
Quantity:
10 g
25 g

Trifluoroacetic acid-d is employed as solvent to investigate the conformations of cyclic peptides by proton NMR spectra. It is also used to compose NMR solvent mixture to investigate the NMR spectra of high molecular weight polyesters such as polyglycolide (PG) and poly(ethylene terephthalate)and as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-00-8
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 115.029
MDL Number MFCD00044563
InChI Key DTQVDTLACAAQTR-DYCDLGHISA-N
Synonym trifluoroacetic acid-d, deuterotrifluoroacetic acid, acetic acid-d, trifluoro, acetic acid-d,2,2,2-trifluoro, acetic acid-d, 2,2,2-trifluoro, trifluoroacetic acid-d tfa-d, trifluoroacetic acid-d, 99.5 atom % d, tfa-d, cf3cood, trifluoroacetic 2h acid
PubChem CID 71502
IUPAC Name deuterio 2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)O

Specifications

Physical Form Liquid
Density 1.50 g/mL
Odor Acrid
Packaging Prescored Ampule
Sensitivity Hygroscopic
Quantity 25 g
Melting Point -15°C
Boiling Point 72°C
UN Number UN2699
Solubility Information Soluble in water.
Formula Weight 115.03
Percent Purity 99.5%
Chemical Name or Material Trifluoroacetic acid-d, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'