missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, sodium salt, 97%

Product Code. p-41283 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10784641

Brand: Thermo Scientific Acros 157930250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2923-18-4 | C2F3NaO2 | 136.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2923-18-4
Molecular Formula C2F3NaO2
Molecular Weight (g/mol) 136.01
MDL Number MFCD00013217
InChI Key UYCAUPASBSROMS-UHFFFAOYSA-M
Synonym sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt
PubChem CID 517019
IUPAC Name sodium;2,2,2-trifluoroacetate
SMILES [Na+].[O-]C(=O)C(F)(F)F

Specifications

Melting Point 207.0°C to 209.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Glass bottle
Linear Formula CF3CO2Na
Quantity 25 g
Beilstein 02, II, 186
Solubility Information Solubility in water: 625g/L water (25°C). Other solubilities: 131g/L in cf3cooh (30°C)
Formula Weight 136.01
Percent Purity 0.97
Physical Form Crystalline Powder
Chemical Name or Material Trifluoroacetic acid, sodium salt, 0.97
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid, sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.